missing translation for 'onlineSavingsMsg'
Learn More

3-Iodooxetane 95.0+%, TCI America™

Catalog No. I08721G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08721G Supplier TCI America Supplier No. I08721G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26272-85-5
Molecular Formula C3H5IO
Molecular Weight (g/mol) 183.98
MDL Number MFCD08544403
InChI Key KBEIFKMKVCDETC-UHFFFAOYSA-N
Synonym 3-iodo-oxetane, 1,3-epoxy-2-iodopropane, oxetane, 3-iodo, 3-iodanyloxetane, 3-iodooxetane, acmc-1cgai, 2-iodo-1,3-epoxypropane, 1,3-?epoxy-?2-?iodopropane
PubChem CID 13356686
IUPAC Name 3-iodooxetane
SMILES IC1COC1

Specifications

Color Yellow
Boiling Point 159°C
Quantity 1 g
UN Number 1993
Formula Weight 183.98
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Iodooxetane
TSCA No
Recommended Storage Refrigerator