missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzotrifluoride, 98+%

Catalog No. AAA1184330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1184330 250 g
AAA1184309 10 g
AAA1184318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1184330 Supplier Thermo Scientific Chemicals Supplier No. A1184330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 401-81-0 | C7H4F3I | 272.009 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401-81-0
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00001049
InChI Key IGISPMBUGPHLBY-UHFFFAOYSA-N
Synonym 1-iodo-3-trifluoromethyl benzene, 3-iodobenzotrifluoride, m-iodobenzotrifluoride, benzene, 1-iodo-3-trifluoromethyl, 3-iodobenztrifluoride, m-trifluoromethyl iodobenzene, 3-iodo-alpha,alpha,alpha-trifluorotoluene, 3-trifluoromethyliodobenzene, 3-trifluoromethyl-iodobenzene, alpha,alpha,alpha-trifluoro-m-iodotoluene
PubChem CID 67868
IUPAC Name 1-iodo-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)I)C(F)(F)F

Specifications

Melting Point -8°C
Density 1.882
Boiling Point 81°C to 82°C (24 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.5185
Quantity 250 g
Beilstein 2088596
Sensitivity Light sensitive
Formula Weight 272.01
Percent Purity ≥98%
Chemical Name or Material 3-Iodobenzotrifluoride
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-934-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only