missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzonitrile 98.0+%, TCI America™

Catalog No. I06605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06605G 5 g
I066025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06605G Supplier TCI America Supplier No. I06605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69113-59-3
Molecular Formula C7H4IN
Molecular Weight (g/mol) 229.02
MDL Number MFCD00079762
InChI Key BGARPMGQRREXLN-UHFFFAOYSA-N
Synonym m-iodobenzonitrile, 3-iodo-benzonitrile, benzonitrile, 3-iodo, 1-cyano-3-iodobenzene, 3-iodobenzenecarbonitrile, pubchem3935, 3-cyanoiodobenzene, 3-iodobenzonitrile, ksc358g7h, acmc-1b930
PubChem CID 144341
IUPAC Name 3-iodobenzonitrile
SMILES C1=CC(=CC(=C1)I)C#N

Specifications

Melting Point 41°C
Quantity 5 g
UN Number 3439
Formula Weight 229.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodobenzonitrile
TSCA No
Recommended Storage Refrigerator