missing translation for 'onlineSavingsMsg'
Learn More

3-Iodoaniline 99.0+%, TCI America™

Catalog No. I00475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00475G 5 g
I004725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00475G Supplier TCI America Supplier No. I00475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-01-7
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.025
MDL Number MFCD00007781
InChI Key FFCSRWGYGMRBGD-UHFFFAOYSA-N
Synonym m-iodoaniline, benzenamine, 3-iodo, aniline, m-iodo, m-aminoiodobenzene, 3-iodobenzenamine, 1-amino-3-iodobenzene, 3-iodo-phenylamine, 3iodoaniline, 3-iodo-aniline, 3-iodophenylamine
PubChem CID 12271
IUPAC Name 3-iodoaniline
SMILES C1=CC(=CC(=C1)I)N

Specifications

Color White-Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 219.03
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Iodoaniline
RTECSNumber BY3750000
TSCA Yes