missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-p-toluic Acid 98.0+%, TCI America™
Supplier: TCI America I070725G
Specifications
| 3-Iodo-p-toluic Acid | |
| 209°C | |
| MFCD00010392 | |
| 3-iodo-4-methyl-benzoic acid, benzoic acid, 3-iodo-4-methyl, zlchem 953, 3-iodo-p-toluicacid, pubchem3947, 4-carboxy-2-iodotoluene, akos bbb/227, ksc449e9t, 3-iodo-4-methyl benzoic acid, paragos 440021 | |
| CC1=C(C=C(C=C1)C(=O)O)I | |
| 262.046 | |
| 262.05 | |
| Crystalline Powder |
| 82998-57-0 | |
| C8H7IO2 | |
| 25 g | |
| LDDHMKANNXWUAK-UHFFFAOYSA-N | |
| 3-iodo-4-methylbenzoic acid | |
| 621640 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 82998-57-0 | |
| 262.046 | |
| LDDHMKANNXWUAK-UHFFFAOYSA-N | |
| 621640 | |
| CC1=C(C=C(C=C1)C(=O)O)I |
| C8H7IO2 | |
| MFCD00010392 | |
| 3-iodo-4-methyl-benzoic acid, benzoic acid, 3-iodo-4-methyl, zlchem 953, 3-iodo-p-toluicacid, pubchem3947, 4-carboxy-2-iodotoluene, akos bbb/227, ksc449e9t, 3-iodo-4-methyl benzoic acid, paragos 440021 | |
| 3-iodo-4-methylbenzoic acid |
Safety and Handling
TSCA : No