missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-p-toluic Acid 98.0+%, TCI America™

Catalog No. p-7131554
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07075G 5 g
I070725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07075G Supplier TCI America Supplier No. I07075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82998-57-0
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00010392
InChI Key LDDHMKANNXWUAK-UHFFFAOYSA-N
Synonym 3-iodo-4-methyl-benzoic acid, benzoic acid, 3-iodo-4-methyl, zlchem 953, 3-iodo-p-toluicacid, pubchem3947, 4-carboxy-2-iodotoluene, akos bbb/227, ksc449e9t, 3-iodo-4-methyl benzoic acid, paragos 440021
PubChem CID 621640
IUPAC Name 3-iodo-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)I

Specifications

Melting Point 209°C
Quantity 5 g
Formula Weight 262.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-p-toluic Acid