CAS RN 82998-57-0
IUPAC Name:
3-iodo-4-methoxybenzoate
Synonyms:
3-iodo-4-methoxybenzoate
Molecular Weight (g/mol):
277.04
Molecular Formula:
C8H6IO3
InChi Key:
HNJSYSWRPCCSQI-UHFFFAOYSA-M
SMILES:
COC1=CC=C(C=C1I)C([O-])=O
1
–
1
of
1
results
1
3-Iodo-p-toluic Acid 98.0+%, TCI America™
CAS: 82998-57-0 Molecular Formula: C8H7IO2 Molecular Weight (g/mol): 262.046 MDL Number: MFCD00010392 InChI Key: LDDHMKANNXWUAK-UHFFFAOYSA-N Synonym: 3-iodo-4-methyl-benzoic acid,benzoic acid, 3-iodo-4-methyl,zlchem 953,3-iodo-p-toluicacid,pubchem3947,4-carboxy-2-iodotoluene,akos bbb/227,ksc449e9t,3-iodo-4-methyl benzoic acid,paragos 440021 PubChem CID: 621640 IUPAC Name: 3-iodo-4-methylbenzoic acid SMILES: CC1=C(C=C(C=C1)C(=O)O)I