missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 98.0+%, TCI America™

Catalog No. I09415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09415G 5 g
I09411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09415G Supplier TCI America Supplier No. I09415G

Chemical Identifiers

CAS 151266-23-8
Molecular Formula C5H4IN5
Molecular Weight (g/mol) 261.03
MDL Number MFCD03787931
InChI Key HQAIUXZORKJOJY-UHFFFAOYSA-N
PubChem CID 1519489
IUPAC Name 3-iodo-2H-pyrazolo[3,4-d]pyrimidin-4-amine
SMILES NC1=NC=NC2=NNC(I)=C12

Specifications

Color Yellow
Quantity 5 g
Synonym 3-iodo-1h-pyrazolo 3,4-d pyrimidin-4-amine, 4-amino-3-iodo-1h-pyrazolo 3,4-d pyrimidine, 4-amino-3-iodopyrazolo 3,4-d pyrimidine, 3-iodo-1h-pyrazolo 3,4-d pyrimidin-4-ylamine, 3-iodo-2h-pyrazolo 3,4-d pyrimidin-4-amine, 1h-pyrazolo 3,4-d pyrimidin-4-amine, 3-iodo, pubchem16192, 1h-pyrazolo 3,4-d pyrimidin-4-amine,3-iodo, 3-iodo-1h-pyrazolo 3,4-d pyrimidine-4-amine
Formula Weight 261.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-1H-pyrazolo[3,4-d]pyrimidin-4-amine