missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-1-(phenylsulfonyl)indole, 95%
CAS: 80360-14-1 | C14H10INO2S | 383.20 g/mol
$116.51 - $257.74
Chemical Identifiers
| CAS | 80360-14-1 |
|---|---|
| Molecular Formula | C14H10INO2S |
| Molecular Weight (g/mol) | 383.20 |
| MDL Number | MFCD09037474 |
| InChI Key | GKYWOZYEMLEJFK-UHFFFAOYSA-N |
| Synonym | 3-iodo-1-phenylsulfonyl-1h-indole, 3-iodo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-iodo-1h-indole, 1-benzenesulfonyl-3-iodoindole, 1-benzenesulfonyl-3-iodo-indole, n-benzenesulfonyl-3-iodoindole, 1-phenylsulfonyl-3-iodo-1h-indole, 1h-indole,3-iodo-1-phenylsulfonyl |
| PubChem CID | 10927077 |
| IUPAC Name | 1-(benzenesulfonyl)-3-iodoindole |
| SMILES | IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH27696MD
|
Thermo Scientific Chemicals
H27696MD |
250 mg |
Each for $116.51
|
|
|||||
|
AAH2769603
|
Thermo Scientific Chemicals
H2769603 |
1 g |
Each for $257.74
|
|
|||||
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Light sensitive. Store in dark place. Incompatible with oxidizing agents.
Chemical Identifiers
| 80360-14-1 | |
| 383.20 | |
| GKYWOZYEMLEJFK-UHFFFAOYSA-N | |
| 10927077 | |
| IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1 |
| C14H10INO2S | |
| MFCD09037474 | |
| 3-iodo-1-phenylsulfonyl-1h-indole, 3-iodo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-iodo-1h-indole, 1-benzenesulfonyl-3-iodoindole, 1-benzenesulfonyl-3-iodo-indole, n-benzenesulfonyl-3-iodoindole, 1-phenylsulfonyl-3-iodo-1h-indole, 1h-indole,3-iodo-1-phenylsulfonyl | |
| 1-(benzenesulfonyl)-3-iodoindole |
Specifications
| 80360-14-1 | |
| C14H10INO2S | |
| 250 mg | |
| 3-iodo-1-phenylsulfonyl-1h-indole, 3-iodo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-iodo-1h-indole, 1-benzenesulfonyl-3-iodoindole, 1-benzenesulfonyl-3-iodo-indole, n-benzenesulfonyl-3-iodoindole, 1-phenylsulfonyl-3-iodo-1h-indole, 1h-indole,3-iodo-1-phenylsulfonyl | |
| GKYWOZYEMLEJFK-UHFFFAOYSA-N | |
| 1-(benzenesulfonyl)-3-iodoindole | |
| 10927077 | |
| 95% |
| 125°C to 129°C | |
| MFCD09037474 | |
| Light sensitive | |
| Insoluble in water. | |
| IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1 | |
| 383.20 | |
| 383.2 | |
| 3-Iodo-1-(phenylsulfonyl)indole |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only