missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1-(phenylsulfonyl)indole, 95%

Catalog No. aah2769603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2769603 1 g
AAH27696MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2769603 Supplier Thermo Scientific Chemicals Supplier No. H2769603

CAS: 80360-14-1 | C14H10INO2S | 383.20 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 80360-14-1
Molecular Formula C14H10INO2S
Molecular Weight (g/mol) 383.20
MDL Number MFCD09037474
InChI Key GKYWOZYEMLEJFK-UHFFFAOYSA-N
Synonym 3-iodo-1-phenylsulfonyl-1h-indole, 3-iodo-1-phenylsulfonyl indole, 1-benzenesulfonyl-3-iodo-1h-indole, 1-benzenesulfonyl-3-iodoindole, 1-benzenesulfonyl-3-iodo-indole, n-benzenesulfonyl-3-iodoindole, 1-phenylsulfonyl-3-iodo-1h-indole, 1h-indole,3-iodo-1-phenylsulfonyl
PubChem CID 10927077
IUPAC Name 1-(benzenesulfonyl)-3-iodoindole
SMILES IC1=CN(C2=CC=CC=C12)S(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 125°C to 129°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 383.2
Percent Purity 95%
Chemical Name or Material 3-Iodo-1-(phenylsulfonyl)indole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only