missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-1-methyl-1H-pyrazole, ≥97%, Thermo Scientific™

Catalog No. CC48311DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC48311DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC48311DA Supplier Thermo Scientific Chemicals Supplier No. CC48311DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92525-10-5
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.002
MDL Number MFCD08435890
InChI Key SITJXRWLFQGWCB-UHFFFAOYSA-N
PubChem CID 13334057
IUPAC Name 3-iodo-1-methylpyrazole
SMILES CN1C=CC(=N1)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-iodo-1-methyl-1h-pyrazole, 1-methyl-3-iodopyrazole, 1h-pyrazole, 3-iodo-1-methyl, 3-iodo-1-methy-1h-pyrazole, iodo-n-methylpyrazole, 3-idio-1-methylpyrazole, 3-iodanyl-1-methyl-pyrazole, 1h-pyrazole,3-iodo-1-methyl, 3 5-iodopyrazole, 1-me
Formula Weight 208
Percent Purity 97%
Chemical Name or Material 3-Iodo-1-methyl-1H-pyrazole
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.