missing translation for 'onlineSavingsMsg'
Learn More

3-Indoxyl phosphate disodium salt

Catalog No. AAJ60552MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60552MD 250 mg
AAJ6055203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60552MD Supplier Thermo Scientific Chemicals Supplier No. J60552MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A histochemical substrate for alkaline phosphatase | CAS: 3318-43-2 | C8H6NNa2O4P | 257.092 g/mol

A histochemical substrate for alkaline phosphatase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3318-43-2
Molecular Formula C8H6NNa2O4P
Molecular Weight (g/mol) 257.092
MDL Number MFCD00040646
InChI Key WCSKWBKPKXJWEG-UHFFFAOYSA-L
Synonym 3-indoxyl phosphate disodium salt, disodium indol-3-yl phosphate, indol-3-yl sodium phosphate, disodium 1h-indol-3-yl phosphate, unii-p1w2d6403e, indoxylphosphate, disodium salt, sodium 1h-indol-3-yl phosphate, 1h-indol-3-ol, dihydrogen phosphate ester , disodium salt, ip, na2, 3-indoxyl phosphate
PubChem CID 76830
IUPAC Name disodium;1H-indol-3-yl phosphate
SMILES C1=CC=C2C(=C1)C(=CN2)OP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Color White
Quantity 250 mg
Formula Weight 257.09
Physical Form Powder
Chemical Name or Material 3-Indoxyl phosphate disodium salt
EINECSNumber 222-014-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only