missing translation for 'onlineSavingsMsg'
Learn More

3-Indazolinone 98.0+%, TCI America™

Catalog No. I09291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I09291G Supplier TCI America Supplier No. I09291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 7364-25-2
Melting Point 250°C
Molecular Formula C7H6N2O
Quantity 1 g
MDL Number MFCD00005685
Synonym 1h-indazol-3-ol, 3-indazolinone, indazolinone, 1h-indazol-3 2h-one, 3-hydroxy-1h-indazole, 3-hydroxyindazole, 1,2-dihydro-3h-indazol-3-one, 3-oxo-1,2-indazole, 3h-indazol-3-one, 1,2-dihydro, 2h-indazol-3-ol
InChI Key SWEICGMKXPNXNU-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)NN2
IUPAC Name 1,2-dihydroindazol-3-one
Molecular Weight (g/mol) 134.138
PubChem CID 81829
Formula Weight 134.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Indazolinone
Show More Show Less
EINECSNumber (5)-0269
TSCA Yes