missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylurea 97.0+%, TCI America™

Catalog No. H043825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H043825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H043825G Supplier TCI America Supplier No. H043825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 701-82-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007945
InChI Key IPRCBIWIPMJXIK-UHFFFAOYSA-N
Synonym 1-3-hydroxyphenyl urea, m-hydroxyphenylurea, 3-ureidophenol, urea, 3-hydroxyphenyl, 3-hydroxyphenyl urea, urea, m-hydroxyphenyl, urea, n-3-hydroxyphenyl, n-carbamyl-m-aminophenol, n-3-hydroxyphenyl urea, urea,n-3-hydroxyphenyl
PubChem CID 12796
IUPAC Name (3-hydroxyphenyl)urea
SMILES C1=CC(=CC(=C1)O)NC(=O)N

Specifications

Melting Point 180°C
Color Yellow
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyphenylurea
EINECSNumber (3)-2192
TSCA Yes