missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyphenylacetylene, 95+%

Catalog No. AC392410050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392410050 5 g Glass bottle
AC392410010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392410050 Supplier Thermo Scientific Chemicals Supplier No. 392410050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10401-11-3 | C8H6O | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10401-11-3
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00078347
InChI Key AODMJIOEGCBUQL-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci
PubChem CID 139144
IUPAC Name 3-ethynylphenol
SMILES OC1=CC=CC(=C1)C#C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color White to Yellow
Density 1.8030g/mL
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5840 to 1.5880 (20°C, 589nm)
Linear Formula HOC6H4C∽CH
Quantity 5 g
Specific Gravity 1.803
Formula Weight 118.13
Percent Purity 95+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxyphenylacetylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only