missing translation for 'onlineSavingsMsg'
Learn More

3-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H12441G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12441G Supplier TCI America Supplier No. H12441G

Specifications

Chemical Name or Material 3-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 87199-15-3
Melting Point 99°C
Color White-Yellow
Molecular Formula C7H9BO3
Quantity 1 g
MDL Number MFCD01317846
Synonym 3-hydroxymethyl phenylboronic acid, 3-hydroxymethylphenylboronic acid, 3-hydroxymethyl phenyl boronic acid, 3-hydroxymethyl benzeneboronic acid, 3-boronobenzyl alcohol, 3-hydroxymethylphenyl boronic acid, 3-hydroxymethyl phenylboronicacid, 3-hydroxymethyl phenyl boranediol, 3-boronophenyl methanol
InChI Key HGTDLKXUWVKLQX-UHFFFAOYSA-N
SMILES OCC1=CC=CC(=C1)B(O)O
IUPAC Name [3-(hydroxymethyl)phenyl]boronic acid
Molecular Weight (g/mol) 151.96
PubChem CID 2734662
Formula Weight 151.96
Physical Form Crystalline Powder
Show More Show Less