missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyglutaronitrile 95.0+%, TCI America™

Catalog No. H08975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08975G 5 g
H08971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08975G Supplier TCI America Supplier No. H08975G
Only null left

Chemical Identifiers

CAS 13880-89-2
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00671545
InChI Key NMFITULDMUZCQD-UHFFFAOYSA-N
Synonym 1,3-Dicyano-2-propanol
PubChem CID 361762
IUPAC Name 3-hydroxypentanedinitrile
SMILES OC(CC#N)CC#N

Specifications

Quantity 5 g
UN Number 3276
Formula Weight 110.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hydroxyglutaronitrile
TSCA No
Recommended Storage Refrigerator