missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxyflavanone 98.0+%, TCI America™

Catalog No. H10251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10251G Supplier TCI America Supplier No. H10251G
Only null left

Chemical Identifiers

CAS 92496-65-6
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.258
MDL Number MFCD00017738
InChI Key JVSPTYZZNUXJHN-UHFFFAOYSA-N
PubChem CID 3534982
ChEBI CHEBI:48022
IUPAC Name 2-(3-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C1C(OC2=CC=CC=C2C1=O)C3=CC(=CC=C3)O

Specifications

Quantity 1 g
Formula Weight 240.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Hydroxyflavanone