missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzohydrazide 98.0+%, TCI America™

Catalog No. H15205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15205G Supplier TCI America Supplier No. H15205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5818-06-4
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00014759
InChI Key OACGSLLKFCMXSX-UHFFFAOYSA-N
Synonym 3-hydroxybenzhydrazide, 3-hydroxybenzoic hydrazide, m-hydroxybenzohydrazide, m-hydroxybenzhydrazide, 3-hydroxybenzoylhydrazine, benzoylhydrazine, m-hydroxy, 3-???phenol, 3-hydroxy benzhydrazide, 3-hydroxy benzohydrazide, 3-hydroxy-benzohydrazide
PubChem CID 79889
IUPAC Name 3-hydroxybenzohydrazide
SMILES C1=CC(=CC(=C1)O)C(=O)NN

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxybenzohydrazide