missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzhydrazide, 98+%

Catalog No. AAL0435706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0435706 5 g
AAL0435703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0435706 Supplier Thermo Scientific Chemicals Supplier No. L0435706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5818-06-4 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5818-06-4
MDL Number MFCD00014759
ChEBI CHEBI:39411

Specifications

Melting Point 157°C to 159°C
Molecular Formula C17H16Cl2O3
Quantity 5 g
Beilstein 638933
Synonym chloropropylate, chlorpropylat, chlorpropylate, rospin, acaralate, chlormite, gesakur, rospine, rospan, isopropyl 4,4'-dichlorobenzilate
InChI Key AXGUBXVWZBFQGA-UHFFFAOYSA-N
SMILES CC(C)OC(=O)C(C1=CC=C(C=C1)Cl)(C2=CC=C(C=C2)Cl)O
IUPAC Name propan-2-yl 2,2-bis(4-chlorophenyl)-2-hydroxyacetate
Molecular Weight (g/mol) 339.21
PubChem CID 22094
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 3-Hydroxybenzhydrazide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-389-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only