missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzenethiol 98.0+%, TCI America™

Catalog No. H11965G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H11965G Supplier TCI America Supplier No. H11965G

Chemical Identifiers

CAS 40248-84-8
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD01317829
InChI Key DOFIAZGYBIBEGI-UHFFFAOYSA-N
Synonym 3-hydroxythiophenol, 3-mercaptophenol, 3-hydroxybenzenethiol, phenol, 3-mercapto, m-hydroxy thiophenol, 3-hydroxy thiophenol, thioresorcin, 3-hydroxythiophenol 3-mercaptophenol, 3-sulphanylphenol, 3-mercapto phenol
PubChem CID 2736613
IUPAC Name 3-sulfanylphenol
SMILES OC1=CC(S)=CC=C1

Specifications

Melting Point 17°C
Color Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 126.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hydroxybenzenethiol