missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Hydroxyacetophenone 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 94°C |
| Boiling Point | 153°C |
| Quantity | 25 g |
| Formula Weight | 136.15 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3′-Hydroxyacetophenone |
Sécurité et manipulation
| EINECSNumber | (3)-1240 |
| TSCA | Yes |