missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-6-methoxyflavone 98.0+%, TCI America™

Numéro de catalogue. H15225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H15225G 5 g
H15221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H15225G Fournisseur TCI America Code fournisseur H15225G
Il en reste null

Identifiants chimiques

CAS 93176-00-2
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
MDL Number MFCD00017639
InChI Key OGURJSOPVFCIOO-UHFFFAOYSA-N
Synonym 3-hydroxy-6-methoxyflavone, 6-methoxyflavonol, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-one, 3-hydroxy-6-methoxy-2-phenyl-4h-chromen-4-on, 4h-1-benzopyran-4-one, 3-hydroxy-6-methoxy-2-phenyl, 3-hydroxy-6-methoxy-2-phenyl-4h-1-benzopyran-4-one
PubChem CID 688676
IUPAC Name 3-hydroxy-6-methoxy-2-phenylchromen-4-one
SMILES COC1=CC2=C(C=C1)OC(=C(C2=O)O)C3=CC=CC=C3

Spécifications

Melting Point 208°C
Color White-Yellow
Quantity 5 g
Formula Weight 268.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-6-methoxyflavone