missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3'-nitro-2-naphthanilide 95.0+%, TCI America™

Catalog No. H031425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H031425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H031425G Supplier TCI America Supplier No. H031425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-65-9
Molecular Formula C17H12N2O4
Molecular Weight (g/mol) 308.29
MDL Number MFCD00021637
InChI Key YZJSKRBKHCLMQC-UHFFFAOYSA-N
Synonym 2-Hydroxy-3-naphthoic Acid m-Nitroanilide, Naphthol AS-BS, Azoic Coupling Component 17, 3-Hydroxy-N-(3-nitrophenyl)-2-naphthamide
PubChem CID 67277
IUPAC Name 3-hydroxy-N-(3-nitrophenyl)naphthalene-2-carboxamide
SMILES OC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Quantity 25 g
Formula Weight 308.29
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-3′-nitro-2-naphthanilide