missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. H07875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07875G 5 g
H078725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07875G Supplier TCI America Supplier No. H07875G
Only null left

Chemical Identifiers

CAS 874-24-8
Molecular Formula C6H4NO3
Molecular Weight (g/mol) 138.10
MDL Number MFCD00006294
InChI Key BRARRAHGNDUELT-UHFFFAOYSA-M
Synonym 3-hydroxypicolinic acid, 3-hydroxy-2-pyridinecarboxylic acid, 3-hydroxypicolinicacid, 2-pyridinecarboxylic acid, 3-hydroxy, 3-hpa, pyridinecarboxylic acid, hydroxy, 3-hydroxypicolinate, 3-hydroxypyridine-2-carboxylate, hydroxypicolinic acid
PubChem CID 13401
ChEBI CHEBI:64342
IUPAC Name 3-hydroxypyridine-2-carboxylate
SMILES OC1=CC=CN=C1C([O-])=O

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 5 g
Formula Weight 139.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-pyridinecarboxylic Acid