missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2'-methyl-2-naphthanilide 97.0+%, TCI America™

Catalog No. H0315500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0315500G 500 g
H031525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0315500G Supplier TCI America Supplier No. H0315500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-61-5
Molecular Formula C18H15NO2
Molecular Weight (g/mol) 277.323
MDL Number MFCD00021634
InChI Key FBLAHUMENIHUGG-UHFFFAOYSA-N
Synonym 2-Hydroxy-3-naphthoic Acid o-Toluidide, Naphthol AS-D, Azoic Coupling Component 18, 3-Hydroxy-N-(o-tolyl)-2-naphthamide
PubChem CID 67273
IUPAC Name 3-hydroxy-N-(2-methylphenyl)naphthalene-2-carboxamide
SMILES CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2O

Specifications

Melting Point 198°C
Quantity 500 g
UN Number 3077
Formula Weight 277.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-2′-methyl-2-naphthanilide
EINECSNumber (5)-2286
TSCA Yes