missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-2-one 95.0+%, TCI America™

Catalog No. H07415ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07415ML 5 mL
H07411ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07415ML Supplier TCI America Supplier No. H07415ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1679-36-3
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00041627
InChI Key LTAPKZGQTMVYMX-UHFFFAOYSA-N
Synonym 3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
PubChem CID 137151
IUPAC Name hex-3-yn-2-one
SMILES CCC#CC(=O)C

Specifications

Boiling Point 77°C
Quantity 5 mL
UN Number 1992
Formula Weight 96.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexyn-2-one