missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-2-un 95,0+%, TCI America™

Numéro de catalogue. H07411ML
Modifier l'affichage
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H07411ML 1 mL
H07415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H07411ML Fournisseur TCI America Code fournisseur H07411ML

Identifiants chimiques

CAS 1679-36-3
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00041627
InChI Key LTAPKZGQTMVYMX-UHFFFAOYSA-N
Synonym 3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
PubChem CID 137151
IUPAC Name hex-3-yn-2-one
SMILES CCC#CC(=O)C

Spécifications

Boiling Point 77°C
Quantity 1 mL
UN Number 1992
Formula Weight 96.13
Percent Purity ≥95.0% (GC)
Physical Form Liquide
Chemical Name or Material 3-Hexyn-2-one