missing translation for 'onlineSavingsMsg'
Learn More

3-Hexanol, 98%

Catalog No. AAB2021022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2021022 100 g
AAB2021014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2021022 Supplier Thermo Scientific Chemicals Supplier No. B2021022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 623-37-0 | C6H14O | 102.177 g/mol

It finds it application as a flavoring agent or adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a flavoring agent or adjuvant.

Solubility
Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 623-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004582
InChI Key ZOCHHNOQQHDWHG-UHFFFAOYSA-N
Synonym 3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
PubChem CID 12178
IUPAC Name hexan-3-ol
SMILES CCCC(CC)O

Specifications

Density 0.81
Boiling Point 134°C to 136°C
Flash Point 42°C (108°F)
Odor Characteristic
Refractive Index 1.415
Quantity 100 g
UN Number UN2282
Beilstein 1718964
Solubility Information Soluble in alcohol.
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material 3-Hexanol
Show More Show Less
Hazard Category H226-H302-H319
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
EINECSNumber 210-790-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only