missing translation for 'onlineSavingsMsg'
Learn More

3-Hexadecanone 98.0+%, TCI America™

Catalog No. H05655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05655G 5 g
H056525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05655G Supplier TCI America Supplier No. H05655G

Chemical Identifiers

CAS 18787-64-9
Molecular Formula C16H32O
Molecular Weight (g/mol) 240.431
MDL Number MFCD00048458
InChI Key LTMXHUUHBSCKEK-UHFFFAOYSA-N
Synonym 3-hexadecanone, ethyl n-tridecyl ketone, ethyl tridecyl ketone, acmc-209eqe, oo>>ue r eyie>>u 1/4 xi feminine
PubChem CID 87789
IUPAC Name hexadecan-3-one
SMILES CCCCCCCCCCCCCC(=O)CC

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 240.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hexadecanone