missing translation for 'onlineSavingsMsg'
Learn More

3-Guanidinopropionic acid, 97%

Catalog No. AC426660010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC426660010 1 g
AC426660050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426660010 Supplier Thermo Scientific Chemicals Supplier No. 426660010

CAS: 353-09-3 | C4H9N3O2 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 353-09-3
Molecular Formula C4H9N3O2
Molecular Weight (g/mol) 131.13
MDL Number MFCD00045939
InChI Key KMXXSJLYVJEBHI-UHFFFAOYSA-N
Synonym 3-guanidinopropionic acid, 3-guanidinopropanoic acid, beta-guanidinopropionic acid, guanidinopropionic acid, 3-guanidinopropanoate, 3-carbamimidamidopropanoic acid, beta-gpa, unii-ul1984yrka, beta-guanadinopropionate, n-aminoiminomethyl-beta-alanine
PubChem CID 67701
ChEBI CHEBI:15968
IUPAC Name 3-(diaminomethylideneamino)propanoic acid
SMILES C(CN=C(N)N)C(=O)O

Specifications

Melting Point 222.0°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)2CO2H
Quantity 1 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material 3-Guanidinopropionic acid
Hazard Category GHS Signal Word: Warning
EINECSNumber 206-530-

RUO – Research Use Only