missing translation for 'onlineSavingsMsg'
Learn More

3-Glycidyloxypropyl(dimethoxy)methylsilane 96.0+%, TCI America™

Numéro de catalogue. G026125G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
G026125G 25 g
G0261500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. G026125G Fournisseur TCI America Code fournisseur G026125G
Il en reste null

Identifiants chimiques

CAS 65799-47-5
Molecular Formula C9H20O4Si
Molecular Weight (g/mol) 220.34
MDL Number MFCD00271003
InChI Key WHGNXNCOTZPEEK-UHFFFAOYSA-N
Synonym 3-[Dimethoxy(methyl)silyl]propyl Glycidyl Ether
PubChem CID 2759249
IUPAC Name dimethoxy-methyl-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](C)(CCCOCC1CO1)OC

Spécifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
Formula Weight 220.34
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Glycidyloxypropyl(dimethoxy)methylsilane
EINECSNumber (2)-2072
TSCA No