missing translation for 'onlineSavingsMsg'
Learn More

3-Furylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F04381G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04381G Supplier TCI America Supplier No. F04381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Furylboronic Acid (contains varying amounts of Anhydride)
CAS 55552-70-0
Color White-Yellow
Molecular Formula C4H5BO3
Quantity 1 g
MDL Number MFCD01319007
Synonym furan-3-boronic acid, 3-furanboronic acid, 3-furylboronic acid, furan-3-yl boronic acid, 3-furanylboronic acid, furan-3-ylboranediol, 3-furan boronic acid, furan-3-yl-3-boronic acid, furan-3-yl-boronic acid, boronic acid, 3-furanyl
InChI Key CYEFKCRAAGLNHW-UHFFFAOYSA-N
SMILES OB(O)C1=COC=C1
IUPAC Name (furan-3-yl)boronic acid
Molecular Weight (g/mol) 111.89
PubChem CID 2734358
Formula Weight 111.89
Physical Form Crystalline Powder
Show More Show Less