missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine-2-carboxaldehyde, 97%, Thermo Scientific™

Numéro de catalogue. p-3485480
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
10g
Emballage:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC30904CB 250mg Amber Glass Bottle
CC30904DA 1g Amber Glass Bottle
CC30904EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC30904CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC30904CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 31224-43-8
Formule moléculaire C6H4FNO
Poids moléculaire (g/mol) 125.102
Numéro MDL MFCD07781234
Clé InChI OZIMPUNGBUYCSP-UHFFFAOYSA-N
Synonyme 3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
PubChem CID 11344017
Nom de l’IUPAC 3-fluoropyridine-2-carbaldehyde
SOURIRES C1=CC(=C(N=C1)C=O)F

Spécifications

Nom ou substance chimique 3-Fluoropyridine-2-carboxaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 125.1
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.