missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F04045G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F04045G Supplier TCI America Supplier No. F04045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Fluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 768-35-4
Molecular Formula C6H6BFO2
Quantity 5 g
MDL Number MFCD00236042
Synonym 3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid
InChI Key KNXQDJCZSVHEIW-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC(F)=C1
IUPAC Name (3-fluorophenyl)boronic acid
Molecular Weight (g/mol) 139.92
PubChem CID 2733986
Formula Weight 139.92
Physical Form Crystalline Powder
Show More Show Less