missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl Acetate 98.0+%, TCI America™

$72.77 - $240.84

Chemical Identifiers

CAS 701-83-7
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00142578
InChI Key IAWZWMGUTKRLQB-UHFFFAOYSA-N
Synonym 1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
PubChem CID 136542
IUPAC Name (3-fluorophenyl) acetate
SMILES CC(=O)OC1=CC(=CC=C1)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F08005G
View Documents
TCI America
F08005G
5 g
Each for $72.77
Only null left
 
F080025G
View Documents
TCI America
F080025G
25 g
Each for $240.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

701-83-7
154.14
IAWZWMGUTKRLQB-UHFFFAOYSA-N
136542
CC(=O)OC1=CC(=CC=C1)F
C8H7FO2
MFCD00142578
1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
(3-fluorophenyl) acetate

Specifications

701-83-7
78°C
MFCD00142578
1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
CC(=O)OC1=CC(=CC=C1)F
154.14
154.14
Liquid
Yellow
C8H7FO2
5 g
IAWZWMGUTKRLQB-UHFFFAOYSA-N
(3-fluorophenyl) acetate
136542
≥98.0% (GC)
3-Fluorophenyl Acetate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'