missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorophenyl Acetate 98.0+%, TCI America™
Supplier: TCI America F080025G
Specifications
| 3-Fluorophenyl Acetate | |
| Yellow | |
| C8H7FO2 | |
| 25 g | |
| IAWZWMGUTKRLQB-UHFFFAOYSA-N | |
| (3-fluorophenyl) acetate | |
| 136542 | |
| ≥98.0% (GC) |
| 701-83-7 | |
| 78°C | |
| MFCD00142578 | |
| 1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa | |
| CC(=O)OC1=CC(=CC=C1)F | |
| 154.14 | |
| 154.14 | |
| Liquid |
Chemical Identifiers
| 701-83-7 | |
| 154.14 | |
| IAWZWMGUTKRLQB-UHFFFAOYSA-N | |
| 136542 | |
| CC(=O)OC1=CC(=CC=C1)F |
| C8H7FO2 | |
| MFCD00142578 | |
| 1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa | |
| (3-fluorophenyl) acetate |
Safety and Handling
TSCA : No