missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorocatechol, 99%

Catalog No. AAB2214506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2214506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2214506 Supplier Thermo Scientific Chemicals Supplier No. B2214506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 363-52-0 | C6H5FO2 | 128.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 363-52-0
Molecular Formula C6H5FO2
Molecular Weight (g/mol) 128.10
MDL Number MFCD00042582
InChI Key DXOSJQLIRGXWCF-UHFFFAOYSA-N
Synonym 3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
PubChem CID 67764
ChEBI CHEBI:39876
IUPAC Name 3-fluorobenzene-1,2-diol
SMILES OC1=CC=CC(F)=C1O

Specifications

Melting Point 71°C to 73°C
Quantity 5 g
UN Number UN1759
Beilstein 2436074
Sensitivity Light sensitive
Formula Weight 128.1
Percent Purity 99%
Chemical Name or Material 3-Fluorocatechol
Hazard Category H302-H314-H334-H335
Hazard Statement GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only