Learn More
3-Fluorobenzenesulfonamide, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 437840010
| Quantity | 1g |
|---|
Description
Identifiants chimiques
| 1524-40-9 | |
| 175.18 | |
| 3-fluorobenzene-1-sulfonamide, benzenesulfonamide, 3-fluoro, 3-fluorobenzenesulphonamide, 3-fbsa, 2weo, pubchem11762, acmc-20a5kf, 3-fluoranylbenzenesulfonamide, ksc180k2p, 3-fluorobenzenesulfonamide | |
| 3-fluorobenzenesulfonamide |
| C6H6FNO2S | |
| CRINBBOGNYCAOV-UHFFFAOYSA-N | |
| 150924 | |
| C1=CC(=CC(=C1)S(=O)(=O)N)F |
Spécifications
| 3-Fluorobenzenesulfonamide | |
| 96.0 | |
| Authentic | |
| Glass Bottle | |
| FC6H4SO2NH2 | |
| 3-fluorobenzene-1-sulfonamide, benzenesulfonamide, 3-fluoro, 3-fluorobenzenesulphonamide, 3-fbsa, 2weo, pubchem11762, acmc-20a5kf, 3-fluoranylbenzenesulfonamide, ksc180k2p, 3-fluorobenzenesulfonamide | |
| C1=CC(=CC(=C1)S(=O)(=O)N)F | |
| 175.18 | |
| 175.18 |
| 1524-40-9 | |
| 100.0 | |
| 96% min. (HPLC) | |
| C6H6FNO2S | |
| 1g | |
| CRINBBOGNYCAOV-UHFFFAOYSA-N | |
| 3-fluorobenzenesulfonamide | |
| 150924 | |
| 97% |
Sécurité et manipulation
GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Promotions