missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzeneboronic acid, 97%

Catalog No. AAB2124714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2124714 25 g
AAB2124703 1 g
AAB2124706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2124714 Supplier Thermo Scientific Chemicals Supplier No. B2124714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 768-35-4 | C6H6BFO2 | 139.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 768-35-4
Molecular Formula C6H6BFO2
Molecular Weight (g/mol) 139.92
MDL Number MFCD00236042
InChI Key KNXQDJCZSVHEIW-UHFFFAOYSA-N
Synonym 3-fluorophenyl boronic acid, 3-fluorobenzeneboronic acid, m-fluorophenylboronic acid, 3-fluoro phenylboric acid, 3-fluorophenyl boranediol, 3-fluorophenylbornic acid, fpba, boronic acid, b-3-fluorophenyl, 3-fluoro-phenyl-boronic acid
PubChem CID 2733986
IUPAC Name (3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC=CC(F)=C1

Specifications

Melting Point 212°C to 215°C
Quantity 25 g
Beilstein 3030632
Formula Weight 139.92
Percent Purity 97%
Chemical Name or Material 3-Fluorobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only