missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroazetidine Hydrochloride 98.0+%, TCI America™

Catalog No. F1186200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1186200MG 200 mg
F11861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1186200MG Supplier TCI America Supplier No. F1186200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 617718-46-4
Molecular Formula C3H7ClFN
Molecular Weight (g/mol) 111.544
MDL Number MFCD05663715
InChI Key PXFUWRWCKSLCLS-UHFFFAOYSA-N
Synonym 3-fluoroazetidine hydrochloride, 3-fluoroazetidine hcl, 3-fluoroazetidinehydrochloride, azetidine, 3-fluoro-, hydrochloride, 3-fluoroazetane hydrochloride, acmc-20dlgf, c3h6fn.hcl, ksc941o8n, 3-fluoro-azetidine hydrochloride, 3-fluoroazetidine hcl salt
PubChem CID 10125054
IUPAC Name 3-fluoroazetidine;hydrochloride
SMILES C1C(CN1)F.Cl

Specifications

Melting Point 130°C
Color White
Quantity 200 mg
Formula Weight 111.54
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoroazetidine Hydrochloride