missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F09541G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09541G Supplier TCI America Supplier No. F09541G

Chemical Identifiers

CAS 871329-82-7
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.919
MDL Number MFCD07363778
InChI Key RMBFBZIEXCTPDB-UHFFFAOYSA-N
Synonym 3-fluoro-5-hydroxyphenyl boronic acid, 3-fluoro-5-hydroxybenzeneboronic acid, 3-fluoro-5-hydroxy-phenyl boronic acid, 3-fluoro-5-hydroxphenyl boronic acid, 3-hydroxyl-5-fluoro-phenyl-boronic acid, acmc-209qgp, 3-fluoro-5-hydroxyphenyl-boronic acid, 3-fluoranyl-5-oxidanyl-phenyl boronic acid, boronic acid,b-3-fluoro-5-hydroxyphenyl
PubChem CID 44717308
IUPAC Name (3-fluoro-5-hydroxyphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)F)O)(O)O

Specifications

Melting Point 62°C
Color White
Quantity 1 g
Formula Weight 155.92
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-5-hydroxyphenylboronic Acid (contains varying amounts of Anhydride)