missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. F10261G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10261G Supplier TCI America Supplier No. F10261G
Only null left

Chemical Identifiers

CAS 719268-92-5
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.054
MDL Number MFCD07780754
InChI Key VFMTUTYBMBTIGA-UHFFFAOYSA-N
Synonym 2-(5-Fluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-Fluoro-3-pyridylboronic Acid Pinacol Ester
PubChem CID 16414268
IUPAC Name 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)F

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine