missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-pyridinecarboxaldehyde 98.0+%, TCI America™

Catalog No. F0975200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0975200MG 200 mg
F09751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0975200MG Supplier TCI America Supplier No. F0975200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31224-43-8
Molecular Formula C6H4FNO
Molecular Weight (g/mol) 125.102
MDL Number MFCD07781234
InChI Key OZIMPUNGBUYCSP-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
PubChem CID 11344017
IUPAC Name 3-fluoropyridine-2-carbaldehyde
SMILES C1=CC(=C(N=C1)C=O)F

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 112°C
Quantity 200 mg
Formula Weight 125.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-2-pyridinecarboxaldehyde