missing translation for 'onlineSavingsMsg'
Learn More

3-Ethynylanisole 97,0+%, TCI America™

Catalog No. E06275G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
E06275G 5 g
E06271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06275G Supplier TCI America Supplier No. E06275G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 768-70-7
Formule moléculaire C9H8O
Poids moléculaire (g/mol) 132.16
Numéro MDL MFCD00160810
Clé InChI ZASXCTCNZKFDTP-UHFFFAOYSA-N
Synonyme 3-ethynylanisole, 3-methoxyphenylacetylene, 1-ethynyl-3-methoxy-benzene, benzene, 1-ethynyl-3-methoxy, 3-ethynyl-1-methoxybenzene, 1-etynyl-3-methoxy-benzene, 3-ethynyl-anisole, m-methoxyphenylacetylene, 3-ethynylanisole 96
PubChem CID 640753
Nom de l’IUPAC 1-ethynyl-3-méthoxybenzène
SOURIRES COC1=CC=CC(=C1)C#C

Specifications

Nom ou substance chimique 3-Ethynylanisole
Couleur Jaune
Point d’ébullition 90 °C
Quantité 5 g
Poids de formule 132.16
Pourcentage de pureté ≥97,0% (GC)
Forme physique Liquide
TSCA Non
Recommended Storage Réfrigérateur