missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methylbenzoxazolium Iodide 98.0+%, TCI America™

Catalog No. E04775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04775G 5 g
E047725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04775G Supplier TCI America Supplier No. E04775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5260-37-7
Molecular Formula C10H12INO
Molecular Weight (g/mol) 289.12
MDL Number MFCD00044141
InChI Key UZWYLWSUBQEUMC-UHFFFAOYSA-M
Synonym 3-ethyl-2-methylbenzoxazolium iodide, 3-ethyl-2-methylbenzo d oxazol-3-ium iodide, 2-methyl-3-ethylbenzoxazolium iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide, benzoxazolium, 3-ethyl-2-methyl-, iodide 1:1, 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide, 3-ethyl-2-methylbenzoxazoliumiodide, acmc-209l0h, ethyl-2-methyl-benzoxazolium iodide, 3-ethyl-2-methylbenzoxazolium iodide, technical grade
PubChem CID 21331
IUPAC Name 3-ethyl-2-methyl-1,3-benzoxazol-3-ium iodide
SMILES [I-].CC[N+]1=C(C)OC2=CC=CC=C12

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 289.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethyl-2-methylbenzoxazolium Iodide
EINECSNumber (5)-0159
RTECSNumber DM5285000
TSCA Yes