missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-iodotoluene, 98%

Catalog No. AAA1083914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1083914 25 g
AAA1083906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1083914 Supplier Thermo Scientific Chemicals Supplier No. A1083914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 175277-95-9 | C9H11I | 246.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175277-95-9
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.091
MDL Number MFCD00052855
InChI Key PCSPENCTMKMGFE-UHFFFAOYSA-N
Synonym 2-ethyl-6-methyliodobenzene, 3-ethyl-2-iodotoluene, acmc-1c540, 1-methyl-2-iodo-3-ethylbenzene, 3-ethyl-2-iodo-1-methylbenzene, benzene, 1-ethyl-3-methyl-2-iodo
PubChem CID 2734195
IUPAC Name 1-ethyl-2-iodo-3-methylbenzene
SMILES CCC1=CC=CC(=C1I)C

Specifications

Boiling Point 118°C to 120°C (15 mmHg)
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 246.08
Percent Purity 98%
Chemical Name or Material 3-Ethyl-2-iodotoluene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only