missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethoxycarbonyl)phenyl isocyanate, 97%

Catalog No. AAL0949514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0949514 25 g
AAL0949503 1 g
AAL0949506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0949514 Supplier Thermo Scientific Chemicals Supplier No. L0949514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67531-68-4 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67531-68-4
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00013865
InChI Key UPXGGIRFDZENRD-UHFFFAOYSA-N
Synonym 3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester
PubChem CID 589806
IUPAC Name ethyl 3-isocyanatobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)N=C=O

Specifications

Density 1.154
Boiling Point 213°C
Flash Point >110°C (230°F)
Refractive Index 1.531
Quantity 25 g
UN Number UN2206
Beilstein 7636626
Sensitivity Moisture sensitive
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material 3-(Ethoxycarbonyl)phenyl isocyanate
Show More Show Less
Hazard Category H302-H315-H317-H319-H331-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only