missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxy-5-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Melting Point | 158°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 183.97 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Ethoxy-5-fluorophenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
| TSCA | No |