missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethoxy-4-methoxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America E083525G
Specifications
| 3-Ethoxy-4-methoxybenzaldehyde | |
| 50°C | |
| 155°C | |
| MFCD00010128 | |
| 3-ethoxy-4-methoxy-benzaldehyde, benzaldehyde, 3-ethoxy-4-methoxy, pubchem13135, 3-ethoxy-p-anisaldehyde, acmc-1ca82, 3-ethoxy-4methoxybenzaldehyde, 3ethoxy-4-methoxybenzaldehyde, ksc183c6j, 3-ethoxy-4-methoxybenzaldehyd, 4-methoxy-3-ethoxybenzaldehyde | |
| CCOC1=C(C=CC(=C1)C=O)OC | |
| 180.203 | |
| 180.20 | |
| Crystalline Powder |
| 1131-52-8 | |
| Yellow | |
| C10H12O3 | |
| 25 g | |
| VAMZHXWLGRQSJS-UHFFFAOYSA-N | |
| 3-ethoxy-4-methoxybenzaldehyde | |
| 244728 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1131-52-8 | |
| 180.203 | |
| VAMZHXWLGRQSJS-UHFFFAOYSA-N | |
| 244728 | |
| CCOC1=C(C=CC(=C1)C=O)OC |
| C10H12O3 | |
| MFCD00010128 | |
| 3-ethoxy-4-methoxy-benzaldehyde, benzaldehyde, 3-ethoxy-4-methoxy, pubchem13135, 3-ethoxy-p-anisaldehyde, acmc-1ca82, 3-ethoxy-4methoxybenzaldehyde, 3ethoxy-4-methoxybenzaldehyde, ksc183c6j, 3-ethoxy-4-methoxybenzaldehyd, 4-methoxy-3-ethoxybenzaldehyde | |
| 3-ethoxy-4-methoxybenzaldehyde |
Safety and Handling
TSCA : No